Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Acetaldehyde ammonia trimer is used as scavengers for sulfhydryl compounds in natural gas.
Solubility
Soluble in water.
Notes
Desiccate at 4°C. Store away from air, moisture and oxidizing agents
RUO – Research Use Only
V.Vinogradoff; F. Duvernay; M. Farabet; G. Danger; P.Theule; F. Borget; J. C. Guillemin and T. Chiavassa. Acetaldehyde Solid State Reactivity at Low Temperature: Formation of the Acetaldehyde Ammonia Trimer. J. Phys. Chem. A.2012, 116, (9), 2225-2233.