Di-p-toluoyl-D-tartaric acid, 98%
Di-p-toluoyl-D-tartaric acid, 98%
Di-p-toluoyl-D-tartaric acid, 98%
Alfa Aesar

Di-p-toluoyl-D-tartaric acid, 98%

CAS: 32634-68-7 | C20H18O8 | 386.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number ALF-L13223-14
HSN 29173990
Price (INR)
5,865.00
Each
Quantity:
25 g
Request bulk or custom format
Price (INR)
5,865.00
Each
Chemical Identifiers
CAS32634-68-7
IUPAC Name(2S,3S)-2,3-bis(4-methylbenzoyloxy)butanedioic acid
Molecular FormulaC20H18O8
InChI KeyCMIBUZBMZCBCAT-HOTGVXAUSA-N
SMILESCC1=CC=C(C=C1)C(=O)O[C@@H]([C@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
CommentSpecification differs for U.S. and non-U.S. material where indicated
Optical Rotation139.4 ± 0.1° (C=1 in ethanol, non-U.S. specification)
Water Content (Karl Fischer Titration)≤3%
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only