RNase-Free Elution Tubes (2.0 mL)
Invitrogen™

RNase-Free Elution Tubes (2.0 mL)

These elution tubes are guaranteed RNase- and DNase-free.
Have Questions?
Catalog NumberQuantity
AM12480100 Tubes
Catalog number AM12480
Price (INR)
14,677.00
Each
Quantity:
100 Tubes
Price (INR)
14,677.00
Each
Invitrogen microcentrifuge tubes are guaranteed RNase- and DNase-free.

Features include:

  • Marked with graduations for convenience
  • Flat lids for easy labeling
  • Positive seal design that prevents caps from opening in boiling water baths
  • Autoclave-safe
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorClear
Dnase FreeRNase-Free, DNase-Free
Product LineInvitrogen
Product TypeElution Tube
Quantity100 Tubes
Shipping ConditionRoom Temperature
Surface TypeGraduated
AutoclavableAutoclavable
Capacity (Metric)2 mL
Closure TypeSnap Cap
GraduatedYes
MaterialPolypropylene
Unit SizeEach
Contents & Storage
Store at room temperature.

Frequently asked questions (FAQs)

Can the collection tubes within the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908) be ordered separately?

Yes, the Elution Tubes (2.0 mL) (Cat. No. AM12480) are the same that are used in the MEGAclear Transcription Clean-Up Kit (Cat. No. AM1908).