Ambion™ RNase-free Conical Tubes are provided assembled with green screw caps and guaranteed RNase- and DNase-free. The tubes have aRead more
Have Questions?
Catalog Number
Quantity
AM12501
200 tubes
Catalog number AM12501
Price (INR)
13,084.00
Each
Quantity:
200 tubes
Price (INR)
13,084.00
Each
Ambion™ RNase-free Conical Tubes are provided assembled with green screw caps and guaranteed RNase- and DNase-free. The tubes have a printed writing space and volume marks on the sidewalls for convenience. These polypropylene tubes are sterile, nuclease-free, nonpyrogenic, and can be autoclaved to 121°C. 200 50-mL tubes are provided.
Each lot of tubes is certified RNase- and DNase-free.
For Research Use Only. Not for use in diagnostic procedures.