Barrier (Filter) Tips, 10 μL size (compatible with Pipetman™ pipettors)
Invitrogen™

Barrier (Filter) Tips, 10 μL size (compatible with Pipetman™ pipettors)

Ambion® Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and areRead more
Have Questions?
Catalog NumberQuantity
AM1264010 racks
Catalog number AM12640
Price (INR)
19,472.00
Online offer
Ends: 31-Dec-2026
32,452.00
Save 12,980.00 (40%)
10 racks
Quantity:
10 racks
Price (INR)
19,472.00
Online offer
Ends: 31-Dec-2026
32,452.00
Save 12,980.00 (40%)
10 racks
Ambion® Barrier tips prevent cross-contamination of samples by means of an aerosol filter. They are RNase- and DNase-free and are ideal for handling RNA, PCR applications, and for working with radioisotopes or toxic chemicals. Ten racks containing 8 x 12 tips each are provided. They are compatible with Pipetman™ pipettors. Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 racks
Shipping ConditionRoom Temperature
FilteredFiltered
For Use With (Equipment)Gilson Pipetman
FormatRack
Product LineInvitrogen
SterilitySterile
Tip StyleBarrier
Volume (Metric)10 μL
Unit Size10 racks
Contents & Storage
Store at room temperature.