The Ambion™ MAXIscript™ T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protectionRead more
Have Questions?
Change view
Catalog Number
Product Type
Quantity
AM1314M
T7 In Vitro Transcription Kit with Manual
100 Reactions
AM1312
T7 In Vitro Transcription Kit
30 Reactions
AM1314
T7 In Vitro Transcription Kit
100 Reactions
3 Options
Catalog number AM1314M
Price (INR)
34,771.00
Online offer
Ends: 30-Sep-2026
46,361.00
Save 11,590.00 (25%)
Each
Product Type:
T7 In Vitro Transcription Kit with Manual
Quantity:
100 Reactions
Price (INR)
34,771.00
Online offer
Ends: 30-Sep-2026
46,361.00
Save 11,590.00 (25%)
Each
The Ambion™ MAXIscript™ T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 100 reactions.
Features:
• High yields of high specific activity, full-length RNA probes in just 10 minutes • Ideal probe synthesis kit for nuclease protection assays, northern and Southern blots, and in situ hybridization • Includes TURBO DNase™ • All kits contain quality-tested DNase/RNase-free reagents
The MAXIscript™ In Vitro Transcription Kit synthesizes RNA probes with specific activities reaching 1 × 109 cpm/μg in just 10 minutes. MAXIscript™ Kits are very efficient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them the perfect companion to ribonuclease protection assays. MAXIscript™ Kits are also an excellent choice for the synthesis of nonisotopically labeled probes. Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into the reaction. MAXIscript™ Kits can also be used for the synthesis of modest amounts of unlabeled RNA (2–6 μg/20 μl reaction) of up to 2 kb.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions100 Reactions
Product LineAmbion, MAXIscript
Product TypeT7 In Vitro Transcription Kit with Manual
PromoterT7
Quantity100 Reactions
Starting MaterialPCR Products, Linearized DNA plasmid
Yield2-6 μg
Final Product TypeSynthetic mRNA, mRNA
FormatKit
Sample TypemRNA, Synthetic mRNA
Unit SizeEach
Contents & Storage
T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.