MEGAshortscript™ T7 Transcription Kit
Invitrogen™
MEGAshortscript™ T7 Transcription Kit
The MEGAshortscript™ T7 Kit is designed for high yields of in vitro- transcribed RNA products, in the 20–500 nucleotide range.Read more
Have Questions?
Catalog NumberQuantity
AM135425 Reactions
Catalog number AM1354
Price (INR)
33,378.00
線上優惠
Ends: 31-Dec-2026
47,682.00
Save 14,304.00 (30%)
Each
Quantity:
25 Reactions
Price (INR)
33,378.00
線上優惠
Ends: 31-Dec-2026
47,682.00
Save 14,304.00 (30%)
Each
The MEGAshortscript™ T7 Kit is designed for high yields of in vitro- transcribed RNA products, in the 20–500 nucleotide range. Using conventional methods, transcription of small templates often results in low mass yields of RNA. Short transcripts require many more transcription initiation events to synthesize a given mass of product. Since the initiation step is the rate-limiting step in transcription reactions, the yield of product from short templates is often low. The MEGAshortscript™ T7 Kit has been designed to overcome the drawbacks involved in conventional in vitro transcription to maximize the yield of short RNA transcripts.
Advantages of the MEGAshortscript™ Kit:
• Efficiently transcribes small templates (less than 300 bases) with consistently high yields
• Designed for use with plasmid, synthetic oligonucleotide, or PCR product templates
Note:The MEGAshortscript™ T7 Kit is optimized for use with the T7 polymerase included in the kit. Using a different polymerase may result in low yields.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
No. of Reactions25 Reactions
Product LineAmbion, MEGAshortscript
Product TypeT7 In Vitro Transcription kit
PromoterT7
Quantity25 Reactions
Shipping ConditionDry Ice
Starting MaterialPCR Products, Linearized DNA plasmid
Final Product TypeSynthetic mRNA, mRNA
FormatKit
Sample TypemRNA, Synthetic mRNA
Unit SizeEach
Contents & Storage
T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Frequently asked questions (FAQs)
What size template should I use with the MEGAshortscript T7 kit?
The MEGAshortscript Kit is designed to function best with templates that code for RNA transcripts in the 20 to 500 nt range. The kit can be used to produce longer RNA but the Invitrogen MEGAscript Kit is more economical for this purpose.
I am planning on ordering Invitrogen GeneArt CRISPR T7 Strings DNA. What do you recommend I use to in vitro transcribe gRNA?
We recommend using our Invitrogen Megashortscript T7 Transcription Kit in order to produce ready-to-use in vitro transcribed gRNA.