Ambion™ SP6 RNA Polymerase, cloned, 20 U/μL
Invitrogen™
Ambion™ SP6 RNA Polymerase, cloned, 20 U/μL
This cloned high-purity Ambion™ SP6 RNA Polymerase has a high specificity for its promoter. The enzyme will transcribe large amountsRead more
Have Questions?
Catalog NumberQuantity
AM20711,000 units, 20 μL
Catalog number AM2071
Price (INR)
28,825.50
Each
Quantity:
1,000 units, 20 μL
Request bulk or custom format
Price (INR)
28,825.50
Each
This cloned high-purity Ambion™ SP6 RNA Polymerase has a high specificity for its promoter. The enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters. One tube containing 1,000 U at a concentration of 20 U/ μL is provided. Its high purity makes them ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA. The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity. Functionality of enzyme is tested in an in vitro transcription reaction under optimal and limiting UTP concentration.
Unit Definition:
One unit is the amount of SP6 RNA Polymerase required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20 U/μL
Quantity1,000 units, 20 μL
SourceDNA
PolymeraseSP6 RNA Polymerase
Unit SizeEach