A polyamine-based Ambion™ siRNA transfection agent that can be used to transfect cells during subculturing-—saving a full day of valuableRead more
Have Questions?
Change view
Catalog Number
Quantity
Product Type
AM4502M
400 μL
Transfection Reagent with Manual
AM4502
400 μL
Transfection Reagent
AM4503M
1 mL
Transfection Reagent with Manual
AM4503
1 mL
Transfection Reagent
4 Options
Catalog number AM4502M
Price (INR)
64,388.00
Each
Quantity:
400 μL
Product Type:
Transfection Reagent with Manual
Price (INR)
64,388.00
Each
A polyamine-based Ambion™ siRNA transfection agent that can be used to transfect cells during subculturing-—saving a full day of valuable time—without increasing toxicity. It is supplied in one tube containing 1 mL. A successful siRNA experiment requires the appropriate transfection agent for your cells, and one that is optimized for siRNA delivery. The efficiency with which different transfection reagents deliver nucleic acids can vary dramatically with the type of nucleic acid being transfected (RNA vs. DNA, single-stranded vs. double-stranded) and the cells being used.
Accessory Products: siPORT™ NeoFX™ Transfection Agent (SKU#s AM4511 and AM4510), a cationic and neutral lipid mixture, is also available. For testing which transfection agent is best with your system, the Silencer™ siRNA Transfection II Kit (SKU# AM1631) is recommended. It contains the two different transfection agents along with controls for transfection optimization.
For Research Use Only. Not for use in diagnostic procedures.