Product Image
Invitrogen™

Silencer™ Negative Control No. 5 siRNA

Ambion™ Silencer™ Negative Control #5 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences. The controlRead more
Have Questions?
Catalog NumberQuantity
AM46425 nmol
Catalog number AM4642
Price (INR)
12,891.00
Online offer
Ends: 31-Dec-2026
21,484.00
Save 8,593.00 (40%)
Each
Quantity:
5 nmol
Price (INR)
12,891.00
Online offer
Ends: 31-Dec-2026
21,484.00
Save 8,593.00 (40%)
Each
Ambion™ Silencer™ Negative Control #5 siRNA has no significant sequence similarity to mouse, rat, or human gene sequences. The control has also been tested in cell-based screens and proven to have no significant effect on cell proliferation, viability, or morphology. 5 nmol is provided.

• Nontargeting with limited sequence similarity to known genes
• Validated for use in human, mouse, and rat cells
• Functionally proven to have minimal effects on cell proliferation and viability
• HPLC purified, duplexed, and ready to use

Negative control siRNAs are essential for determining transfection efficiency and to control for the effects of siRNA delivery. In siRNA screening applications, negative control siRNAs are also critical for setting the “hit” threshold that determines whether an siRNA is considered to have a positive, negative, or neutral effect in a particular assay.

Accessory Product: Silencer™ siRNA Screening Control Panel (SKU#AM4640)
The Silencer™ siRNA Screening Control Panel includes seven negative control siRNAs and is for those researchers performing siRNA screens that employ hundreds or thousands of siRNAs per experiment. Because negative control siRNAs may have unintended effects in a particular assay, a negative control siRNA should be carefully chosen. The Silencer™ siRNA Screening Control Panel simplifies this process. As an added benefit, a positive control siRNA to human, mouse, and rat KIF11 (Eg5) is included in the panel. Knockdown of KIF11, which encodes a kinesin family motor protein, leads to mitotic arrest. Silencer™ control siRNAs are unmodified 21-mers; they are designed to be used with Silencer™ siRNAs, BLOCK-iT™ siRNAs, and other unmodified 21mer siRNAs. If you are using other siRNA products, such as Ambion™ Silencer™ Select siRNAs or STEALTH RNAi™ siRNAs, we recommend that you use control siRNAs designed for use with those siRNAs for more reliable experimental results.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Transfection, RNAi
Label or DyeUnconjugated
Product LineSilencer, Ambion
Product TypesiRNA
PurityHPLC
Quantity5 nmol
Shipping ConditionDry Ice
Control TypeNegative Control
RNAi TypesiRNA
SpeciesHuman, Mouse, Rat
Unit SizeEach
Contents & Storage
Store at or below -20°C. Do not store in a frost-free freezer.

Frequently asked questions (FAQs)

How critical is it to include positive and negative controls in siRNA experiments?

A positive control siRNA such as the Silencer Select GAPDH siRNA will demonstrate efficient transfection and help to optimize transfection conditions. A negative control siRNA is necessary to serve as a baseline for gene expression.

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.

What are the positive and negative controls I should use with my Silencer or Silencer Select siRNA experiment?

We offer several negative and positive controls for both Silencer and Silencer Select siRNA.

For Silencer siRNA positive control:
- Silencer Firefly Luciferase (GL2 + GL3) siRNA (Cat. No. AM4629, 5nmol)
- Silencer GAPDH siRNA (human) (Cat. No AM4605, 5nmol; AM4633, 40 nmol)
- Silencer GAPDH siRNA (human, mouse, rat) (Cat. No. AM4624, 5 nmol; AM4631, 40 nmol; AM4632, 5 x 40 nmol)
- Silencer GFP (eGFP) siRNA (Cat. No. AM4626, 5 nmol)
- Silencer KIF11 (Eg5) siRNA (human, mouse, rat) (Cat. No. AM4639, 5 nmol)

If working with Fluorescently labeled Silencer siRNA:
- Silencer Cy-3 labeled GAPDH siRNA (human, mouse, rat) (Cat. No. AM4649, 5 nmol)
- Silencer FAM-labeled GAPDH siRNA (human, mouse, rat) (Cat. NO. AM4650, 5 nmol)

If working with Silencer siRNA in vivo ready:
- Silencer GAPDH Positive Control siRNA, in vivo ready (Cat. No. 4404025, 250 nmol)

For Silencer siRNA negative controls:
- Silencer Negative Control No. 1 siRNA (Cat. Nos. AM4611, 5 nmol; AM4635, 40 nmol; AM4636, 5 x 40 nmol)
- Silencer Negative Control No. 2 siRNA (Cat. Nos. AM4613, 5 nmol; AM4637, 30 nmol)
- Silencer Negative Control No. 3 siRNA (Cat. No. AM4615, 5 nmol)
- Silencer Negative Control No. 4 siRNA (Cat. No. AM4641, 5 nmol)
- Silencer Negative Control No. 5 siRNA (Cat. No. AM4642, 5 nmol)

If working with Fluorescently labeled Silencer siRNA:
- Silencer Cy 3-labeled Negative Control No. 1 siRNA (Cat. No AM4621, 5 nmol)
- Silencer FAM-labeled Negative Control No. 1 siRNA (Cat. No. AM4620, 5 nmol)

If working with Silencer siRNA in vivo ready:
- Silencer in vivo ready (Cat. No. 4404021)

For Silencer Select siRNA Positive control:
- Silencer Select GAPDH Positive Control siRNA (Cat. No. 4390849, 5 nmol; 4390850, 40 nmol; 4404024, 250 nmol)
- Silencer Select MALAT1 Positive Control siRNA for non-coding siRNA (Cat. No. 4455877, 5 nmol)

For Silencer Select siRNA negative control:
- Silencer Select Negative Control No. 1 siRNA (Cat. No. 4390843, 5 nmol; 4390844, 40 nmol)
- Silencer Select Negative Control No. 2 siRNA (Cat. No. 4390846, 5 nmol; 4390847, 40 nmol)

If working with Silencer Select siRNA in vivo ready:
- Silencer Select Negative Control No. 1 siRNA, in vivo ready (Cat. No. 4404020, 250nmol)

Find additional tips, troubleshooting help, and resources within our RNAi Support Center.