RNA Control 250 with Manual
Invitrogen™
RNA Control 250 with Manual
The Ambion™ RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM7155M2 x 20 μL, with Manual
AM71552 x 20μL
Catalog number AM7155M
Price (INR)
14,192.00
온라인 행사
Ends: 30-Sep-2026
25,802.00
Save 11,610.00 (45%)
Each
Quantity:
2 x 20 μL, with Manual
Price (INR)
14,192.00
온라인 행사
Ends: 30-Sep-2026
25,802.00
Save 11,610.00 (45%)
Each
The Ambion™ RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers. It is provided in two tubes containing 20 μL each.
• A stable, highly pure transcript at a precisely known concentration
• Used to verify the accuracy of NanoDrop™ Spectrophotometer measurements
• Provided with THE RNA Storage Solution for blank measurements
• Enough for 20 measurements
RNA Control 250 consists of a single, ultra-pure, 2000 nt RNA transcript of a precisely defined sequence, molecular weight, and concentration. It is packaged in special non-stick tubes and includes the Ambion™ THE RNA Storage Solution. The RNA Control 250 will maintain ± 5% of stated concentration and >85% RNA integrity through 5 freeze/thaw cycles (10 duplicate measurements).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
Product LineAmbion
Quantity2 x 20 μL, with Manual
Label or DyeUnconjugated
Unit SizeEach
Contents & Storage
RNA Control and THE RNA Storage Solution should be stored at –20°C
Frequently asked questions (FAQs)
What is the sequence of RNA Control 250?
The sequence of RNA Control 250 is proprietary.