Product Image
Invitrogen™

E. coli Total RNA

The highest-quality RNA available from Ambion®, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 200Read more
Have Questions?
Catalog NumberQuantity
AM7940200 μg
Catalog number AM7940
Price (INR)
-
Quantity:
200 μg
The highest-quality RNA available from Ambion®, it is DNase-treated and subjected to unsurpassed quality control standards. One tube containing 200 μg is provided at a concentration of 1 mg/mL. The samples are processed using Ambion® RNA isolation reagents to produce highly pure, intact RNA. A stringent DNase treatment is performed to ensure that the RNA is ready for use in any downstream application, including RT-PCR. Note: While it is impossible to remove every DNA molecule from an RNA sample, each preparation is tested to ensure that any residual DNA contamination is insignificant. The integrity of the RNA is verified by capillary electrophoresis using the Agilent® 2100 bioanalyzer.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Bacterial ContentRNA: varies
Concentration1 mg/mL
DescriptionE. coli total RNA isolated from DH5α cells
Final Product TypeE. coli total RNA stored in THE RNA Storage Solution (1 mM Sodium Citrate, pH 6.4; Cat No. AM7000)
For Use With (Application)RT-PCR, ribonuclease protection assays, S1 nuclease assays, cDNA synthesis, Northern analysis, IVT
Quantity200 μg
Sample TreatmentDNase-Treated
Shipping ConditionDry Ice
Unit SizeEach
Contents & Storage
200 μg E. coli Total RNA; store at or below –70°C

Frequently asked questions (FAQs)

Do you offer isolated, purified RNA?

Yes, please visit this website (http://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/rna-extraction/rna-types/purified-rna.html) for our FirstChoice total RNA isolated from human tissues, as well as from E. coli and human cell lines.