SSPE (20X), RNase-free
Invitrogen™

SSPE (20X), RNase-free

Molecular biology grade, Ambion® 20X SSPE solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free,Read more
Have Questions?
Catalog NumberQuantity
AM97671 L
Catalog number AM9767
Price (INR)
12,551.00
Each
Quantity:
1 L
Price (INR)
12,551.00
Each
Molecular biology grade, Ambion® 20X SSPE solution is supplied in one bottle containing 1 L. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Ambion®'s nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration20 X
For Use With (Application)Nucleic Acid Gel Electrophoresis & Blotting
FormLiquid
Purity or Quality GradeMolecular Biology Grade
Quantity1 L
Shipping ConditionRoom Temperature
Volume (Metric)1 L
Product LineAmbion
Product TypeSSPE Solution
Unit SizeEach
Contents & Storage
Store at room temperature.