Tris (1 M), pH 8.0, RNase-free
Invitrogen™

Tris (1 M), pH 8.0, RNase-free

Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
AM9855G100 mL
AM9856500 mL
Catalog number AM9855G
Price (INR)
3,179.00
Offre exceptionnelle en ligne
Ends: 31-Dec-2026
5,298.00
Save 2,119.00 (40%)
Each
Quantity:
100 mL
Price (INR)
3,179.00
Offre exceptionnelle en ligne
Ends: 31-Dec-2026
5,298.00
Save 2,119.00 (40%)
Each

Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Formulation1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride
Recommended StorageStore at room temperature.
Concentration1 M
GradeMolecular Biology
Physical FormLiquid
Product LineAmbion
Quantity100 mL
pH8
Unit SizeEach

Frequently asked questions (FAQs)

What alternative do you offer for UltraPure 1M Tris-HCI, pH 8.0 (Cat. No. 15568025)?

We recommend Tris (1 M), pH 8.0, RNase‐free (Cat. No. AM9855G).

Find additional tips, troubleshooting help, and resources within our Nucleic Acid Purification and Analysis Support Center.