EVOS™ 60X Objective, fluorite, LWD
Invitrogen™
EVOS™ 60X Objective, fluorite, LWD
This fluorite objective is ideal for fluorescence and demanding transmitted-light applications. This is a long working distance (LWD) objective thatRead more
Have Questions?
Catalog NumberQuantity
AMEP46261 Ea.
Catalog number AMEP4626
Price (INR)
224,522.00
Precio exclusivo compra web
Ends: 31-Dec-2026
332,625.00
Save 108,103.00 (33%)
Each
Quantity:
1 Ea.
Price (INR)
224,522.00
Precio exclusivo compra web
Ends: 31-Dec-2026
332,625.00
Save 108,103.00 (33%)
Each
This fluorite objective is ideal for fluorescence and demanding transmitted-light applications. This is a long working distance (LWD) objective that is optimized for the imaging of slides, cell culture dishes and flasks, and microtiter plates. All EVOS objectives offer outstanding optical performance from visible light to near infrared light. The extensive choice of objectives satisfies needs across the spectrum of magnifications and optical specifications.
Additional characteristics of this EVOS objective:
• Magnification: 60X
• Numerical Aperture: 0.75
• Working Distance: 2.2 mm
Image Quality
Microscope objectives may be the most important components of an optical microscope because they are responsible for primary image formation. Image quality is crucial to experimental success and a requirement for publication; EVOS objectives afford that quality across the visible spectrum to near infrared light. This performance results from years of lens manufacture perfection. EVOS objectives have the same or better numerical apertures as any other manufacturer's in the same class, and the broad selection means you have choices for your imaging requirements.
Objective Classes
Achromat objectives are perfect for general applications, with standard correction of color and focus.
Fluorite objectives deliver excellent resolution and are made with higher numerical apertures than achromat objectives, resulting in brighter fluorescence signal and higher contrast imaging. The higher optical quality greatly reduces optical aberrations, and corrections for color and focus are at higher levels than achromat objectives. Fluorite objectives are ideally suited for fluorescence and demanding transmitted light applications, where the higher contrast make them ideally suited for color imaging.
Apochromat objectives are manufactured to the highest levels of resolution, fluorescence brightness, and contrast; chromatic aberrations are almost eliminated. They are recommended for the most demanding applications, particularly at magnifications of 60x and above. Apochromatic objectives are the best choice for the capture of color images in white light.
Brightfield Contrast versus Phase Contrast Objectives
Brightfield is the most basic form of light microscopy and is accomplished by sample absorption of light. A higher density area in a sample will absorb more light, thus increasing contrast in those areas.
Phase contrast objectives are most useful for hard to see, translucent specimens. This method of contrast is accomplished by converting phase shifts, caused by light passing through a translucent specimen, into brightness changes (i.e., contrast).
Long Working Distance versus Coverslip-Corrected Objectives
Long working distance (LWD) objectives are optimized for use through vessels with a nominal wall thickness of 0.9-1.5 mm. This includes vessels commonly used in cell culture and cell-based assays, such as slides, cell culture dishes and flasks, microtiter well plates, etc. Coverslip-corrected objectives are optimized for use through #1.5 coverslips (thickness approximately 0.17 mm). These objectives have a higher magnification/NA ratio and provide higher resolution compared to LWD objectives.
For additional choices, visit the EVOS Objectives selection guide
Explore the entire EVOS line of imaging systems and accessories
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
IncludesVessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Lens TypePlan Fluorite
Magnification60X
Numerical Aperture0.75
Quantity1 Ea.
Product LineEVOS
TypeObjective Lens
Unit SizeEach
Frequently asked questions (FAQs)
I'm using an EVOS imaging system and my objective is rubbing up against the edge of the vessel holder of my stage. How can I correct this?
Objectives can hit the vessel holder when they are focused too high in the Z axis (up and down). This is a particularly a problem with the EVOS FL Auto Imaging System during instrument start-up, when the stage moves during system initiation, or when changing objectives. Coverslip-corrected objectives tend to be wider and flatter at the top of the barrel, which means that they are more likely to run into the edges of the vessel holder, particularly if you are imaging at the edges of the sample container. In those cases, use of that objective for those areas of the container may not be possible. If the objective if “jammed” by the vessel holder, then carefully unscrew the thumbscrews of the vessel holder and lift it straight off the stage, then move the objective downward in focus and toward the center of the stage. It is a good idea to have a shut-down procedure in your lab that includes moving the objectives to the lowest magnification and focusing downward with course focus prior to turning off the instrument for the day.
An objective can be damaged by scraping against the vessel holder. If this happens, take out the objective and examine it carefully for damage, particularly on the lens.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
My objective keeps running into my sample when I'm trying to focus with my EVOS FL Auto Imaging System. What can I do about this?
If the lens is running up into a sample, this may be an issue with either focusing too quickly and missing the focal plane (if focusing manually) or a problem with the objective calibration (if using autofocus). It is a good idea to calibrate your objectives using the FL Auto calibration slide that comes with the system. Check to see if your objective is a long-working distance (LWD) or coverslip-corrected objective (CC). If coverslip-corrected, it is only for use with very short working distances for imaging through thin coverslips, but not through the slide or through plastics in microplates or culture dishes). If working with high magnification and oil immersion, by eye, move the objective upwards to touch the bottom of the sample and then only move slowly away from the sample for further focusing. An objective lens can be seriously damaged by scraping against samples. If this happens, check the objective lens for damage.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
The objectives available for the EVOS cell imaging systems come in different formats: Plan Achromat, Plan Fluorite, and Plan Apochromat. What factors should I consider when selecting an objective?
Plan Achromat objectives are perfect for general applications where color and focus have standard correction; these are suitable for samples requiring low magnification (2x to 4x). Plan Achromat objectives are recommended for basic brightfield microscopy and simple fluorescence detection.
Plan Fluorite objectives provide the next level of improved resolution for brighter fluorescence signal and high contrast. These objectives are recommended for basic fluorescence imaging and brightfield microscopy at higher magnifications.
Plan Apochromat provides the highest level of resolution, fluorescence brightness, contrast, and chromatic correction. If you are imaging very small structures and require high contrast and brightness, the Plan Apochromat objectives are the best option.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the difference between "long working distance" (LWD) objectives and "coverslip corrected" (CC) objectives in the EVOS imaging systems?
All the EVOS imaging systems are inverted microscopes. For CC objectives, the coverslip must be face down, facing the objectives as the lenses have a short working distance suitable only for thin glass or plastic coverslips. LWD objectives are designed for viewing from the bottom of microplates, petri dishes, or culture flasks; the longer working distances of the lenses in these objectives accommodate thicker materials such as the plastic bottoms of various vessels.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.