EVOS™ XL Core Attachable Mechanical Stage
EVOS™ XL Core Attachable Mechanical Stage
The EVOS XL Core Mechanical Stage is an alternative to the universal, fixed stage provided with the EVOS XL CoreRead more
Have Questions?
Catalog NumberQuantity
AMEP47121 Ea.
Catalog number AMEP4712
Price (INR)
122,470.00
온라인 행사
Ends: 30-Sep-2026
222,671.00
Save 100,201.00 (45%)
Each
Quantity:
1 Ea.
Price (INR)
122,470.00
온라인 행사
Ends: 30-Sep-2026
222,671.00
Save 100,201.00 (45%)
Each
The EVOS XL Core Mechanical Stage is an alternative to the universal, fixed stage provided with the EVOS XL Core Imaging System. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders for smaller sample vessel types, such as microscope slides and 35 mm dishes. Larger sample vessels, such as T-75 flasks or multi-well plates, fit directly onto the stage. Both vessel holders and large vessels are held securely in place by a stage clip during operation. The spring-loaded clip allows fast and easy vessel changes, such as during operations inside a hood or during processing of multiple samples.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS™ XL Core Imaging System
Quantity1 Ea.
Product LineEVOS
TypeMechanical Stage
Unit SizeEach
Frequently asked questions (FAQs)
What is the EVOS XL Core Attachable Mechanical Stage for?
The EVOS XL Core Attachable Mechanical Stage (Cat. No. AMEP4712) is an optional feature, that can be purchased separately. It is an alternative to the universal, fixed stage provided with the EVOS XL Core Imaging System and allows for easier manipulation of sample vessels and slides. This mechanical stage has X-Y axis controls and a vessel holder framework that accommodates our broad range of vessel holders for smaller sample vessel types, such as microscope slides and 35 mm dishes. Larger sample vessels, such as T-75 flasks or multi-well plates, fit directly onto the stage.