EVOS™ Onstage Incubator Hose
EVOS™ Onstage Incubator Hose
The EVOS Onstage Incubator Hose connects the Control Unit and the Stagetop Environmental Chamber units of the EVOS Onstage IncubatorRead more
Have Questions?
Catalog NumberQuantity
AMEP47281 each
Catalog number AMEP4728
Price (INR)
9,851.00
キャンペーン価格
Ends: 31-Mar-2026
16,418.00
Save 6,567.00 (40%)
Each
Quantity:
1 each
Price (INR)
9,851.00
キャンペーン価格
Ends: 31-Mar-2026
16,418.00
Save 6,567.00 (40%)
Each
The EVOS Onstage Incubator Hose connects the Control Unit and the Stagetop Environmental Chamber units of the EVOS Onstage Incubator and helps precisely control the temperature. Even small temperature shifts can have profound effects on dynamic cellular events. When performing live cell imaging studies, it is essential that the physiological and biological processes under investigation are not altered in any way. The ability to maintain temperature control with a precision of 0.1°C helps maintain cell viability and function and helps reduce the risk of biological temperature-related artifacts.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS™ FL Auto Imaging System
Quantity1 each
Product LineEVOS
TypeOnstage Incubator Hose
Unit SizeEach