EVOS™ M7000 Computer Upgrade
EVOS™ M7000 Computer Upgrade
EVOS™ M7000 Computer Upgrade
Invitrogen™
EVOS™ M7000 Computer Upgrade
AMEP5023 contains a customized Dell PC and EVOS Deconvolution license (AMEP5022).
Have Questions?
Catalog NumberFor Use With (Equipment)
AMEP5023EVOS M7000 Imaging System
Catalog number AMEP5023
Price (INR)
-
For Use With (Equipment):
EVOS M7000 Imaging System
The latest EVOS M7000 Imaging System is powered by an external Dell T2 Pro computer with Intel Core i9 processor. The Dell T2 Pro computer offers increased memory (128 GB DDR5 RAM) and storage (4 TB SSHD) compared to previous computer models, along with similar NVIDIA Quadro RTXA4000 with 16 GB discrete video graphics and Windows 11 LTSC operating system.
If you currently own an EVOS M7000 system with a previous model computer (Dell XE3, Dell 3650, or XE4), then purchase of this product enables an upgrade to the latest computer platform and operating system offered for the EVOS M7000 system, with the benefit of improved processor and increased memory and storage.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS M7000 Imaging System
IncludesDell T2 Pro PC with 2D Deconvolution License Activated
Unit SizeEach