EVOS™ Condenser Slider, Green
EVOS™ Condenser Slider, Green
Condenser sliders are used to improve illumination under certain conditions and several are available for the EVOS imaging systems, includingRead more
Have Questions?
Catalog NumberQuantity
AMEPCFGEach
Catalog number AMEPCFG
Price (INR)
5,432.00
Each
Quantity:
Each
Price (INR)
5,432.00
Each
Condenser sliders are used to improve illumination under certain conditions and several are available for the EVOS imaging systems, including a diffuser slider, pinhole slider, and meniscus sliders. Due to variances in sample size, color and thickness, actual slider use may differ from suggestions. This green slider is intended for brightfield applications at all magnifications to enhance phase-contrast.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
IncludesDiffuser Slider, Pinhole Slider, Meniscus Sliders
QuantityEach
Product LineEVOS
TypeCondenser Slider
Unit SizeEach