EVOS™ Condenser Slider, Meniscus B (semi-opaque liquids)
EVOS™ Condenser Slider, Meniscus B (semi-opaque liquids)
Condenser sliders are used to improve illumination under certain conditions and several are available for the EVOS imaging systems, includingRead more
Have Questions?
Catalog NumberQuantity
AMEPMN41 each
Catalog number AMEPMN4
Price (INR)
3,093.00
Online offer
Ends: 31-Dec-2026
5,155.00
Save 2,062.00 (40%)
Each
Quantity:
1 each
Price (INR)
3,093.00
Online offer
Ends: 31-Dec-2026
5,155.00
Save 2,062.00 (40%)
Each
Condenser sliders are used to improve illumination under certain conditions and several are available for the EVOS imaging systems, including a diffuser slider, pinhole slider, and meniscus sliders. Due to variances in sample size, color and thickness, actual slider use may differ from suggestions. This Meniscus B slider is intended for brightfield applications at 2x magnification with high-volume samples in multi-well plates and with semi-opaque liquids.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
IncludesDiffuser Slider, Pinhole Slider, Meniscus Sliders
Quantity1 each
Product LineEVOS
TypeMeniscus B Condenser Slider
Unit SizeEach