EVOS™ Onstage Master Plate
EVOS™ Onstage Master Plate
The EVOS Onstage Master Plate is part of the EVOS Onstage Incubator and holds the EVOS Onstage Vessel Holder. TheRead more
Have Questions?
Catalog NumberQuantity
AMEPVH0351 Ea.
Catalog number AMEPVH035
Price (INR)
37,767.00
キャンペーン価格
Ends: 30-Sep-2026
68,667.06
Save 30,900.06 (45%)
Each
Quantity:
1 Ea.
Price (INR)
37,767.00
キャンペーン価格
Ends: 30-Sep-2026
68,667.06
Save 30,900.06 (45%)
Each
The EVOS Onstage Master Plate is part of the EVOS Onstage Incubator and holds the EVOS Onstage Vessel Holder. The EVOS Onstage Incubator is an environmental chamber designed specifically for use with the EVOS FL Auto Imaging System. This master plate plus vessel holder design, with vessel holders available that accommodate various types of cell culture vessels, provides flexibility in your approach to extended time-lapse imaging experiments under precisely controlled environmental conditions. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)EVOS™ FL Auto Imaging System
HoldsOnstage Vessel
Quantity1 Ea.
Product LineEVOS
TypeMaster Plate
Unit SizeEach