EVOS™ Stage Plate for Automated Stage, one Nunc T-175 flask
Invitrogen™
EVOS™ Stage Plate for Automated Stage, one Nunc T-175 flask
This EVOS stage plate is intended for use on the stage of the EVOS FL Auto Imaging System. This stageRead more
Have Questions?
Catalog NumberQuantity
AMEPVH0421 plate
Catalog number AMEPVH042
Price (INR)
30,087.00
Online offer
Ends: 30-Sep-2026
40,116.00
Save 10,029.00 (25%)
Each
Quantity:
1 plate
Price (INR)
30,087.00
Online offer
Ends: 30-Sep-2026
40,116.00
Save 10,029.00 (25%)
Each
This EVOS stage plate is intended for use on the stage of the EVOS FL Auto Imaging System. This stage plate is designed to hold one Nunc T-175 flask for increased precision in sample alignment and is just one of many easy-to-change stage plates and vessel holders available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
For additional choices, see the EVOS Guide to Vessel Holders and Stage Plates.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionStage Plate for Automated Stage, one Nunc™ T-175 flask
Dimensions259 mm x 204 mm
For Use With (Equipment)EVOS™ FL Auto Imaging System
Holds1 Nunc T-175 flask
Quantity1 plate
Shipping ConditionRoom Temperature
Product LineEVOS
TypeImaging System Stage Plate
Unit SizeEach