EVOS™ Vessel Holder for Automated Stage, one Falcon T-75 flask or standard micro-well plate
Invitrogen™
EVOS™ Vessel Holder for Automated Stage, one Falcon T-75 flask or standard micro-well plate
EVOS vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate toRead more
Have Questions?
Catalog NumberQuantity
AMEPVH0541 holder
Catalog number AMEPVH054
Price (INR)
19,660.00
キャンペーン価格
Ends: 30-Sep-2026
26,212.72
Save 6,552.72 (25%)
Each
Quantity:
1 holder
Price (INR)
19,660.00
キャンペーン価格
Ends: 30-Sep-2026
26,212.72
Save 6,552.72 (25%)
Each
EVOS vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment. This particular vessel holder securely holds one Falcon T-75 flask or standard micro-well plate and is specific for the automated stage of the EVOS FL Auto Imaging System. It is just one of many easy-to-change vessel holders and stage plates available for this instrument platform. With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
For additional choices, see the EVOS Guide to Vessel Holders and Stage Plates.
Explore the entire EVOS line of imaging systems and accessories.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionVessel Holder for Automated Stage, one Falcon T-75 flask or standard micro-well plate
Dimensions259 mm x 204 mm
For Use With (Equipment)EVOS™ FL Auto Imaging System
Holds1 BD-Falcon T-75 flask, 1 multi-well plate
Quantity1 holder
Shipping ConditionRoom Temperature
Product LineEVOS
TypeImaging System Vessel Holder
Unit SizeEach
Frequently asked questions (FAQs)
How should the EVOS vessel holders be stored?
Store the EVOS vessel holders flat and protected from any contact with heavy objects as they can be distorted. Once the holders are distorted and no longer lay flat on the stage, they cannot be used. Store them individually in their own lidded plastic containers or small sturdy boxes.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
How should I clean my EVOS vessel holders?
Wipe the vessel holder with paper towels or Kimwipes tissues dampened with 70% ethanol or 4,000 ppm hydrogen peroxide (H2O2).
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
I have an EVOS vessel that does not fit into any of the vessel holders that you offer. What are my options?
We do offer an option for making custom vessel holders. Please contact your Sales Representative for initiating a custom order.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.