Antaris™ IGS Gas Cells
Antaris™ IGS Gas Cells
Antaris™ IGS Gas Cells
Thermo Scientific™
Antaris™ IGS Gas Cells
Meet the requirements of today's demanding gas applications with Thermo Scientific™ Antaris™ IGS Gas Cells.
Have Questions?
Catalog number ANTARISIGSGAS
Price (INR)
-
Built to meet the needs of industrial customers, the Thermo Scientific™ Antaris™ IGS Gas Cells are multi-pass cells that provide superior FT-IR gas analysis capabilities. Optimized for integration with the Thermo Scientific™ Antaris™ IGS Gas Analyzer, the Antaris IGS Gas Cells combine industrial design with the precision and configurability necessary for real world gas analysis. They offer a range of sensitivities and chemical compatibilities to meet the requirements of today's demanding gas applications.
The Antaris IGS Gas Cells incorporate high-quality optical components to ensure exceptional results and allow for coverage of a wide variety of samples. These gas cells provide low cost-of-ownership while still offering exceptional sensitivity, especially in physically demanding environments.
The Antaris IGS Gas Cells Feature:
  • Low ppb detection limits for most components
  • Precisely known pathlengths for excellent calibration confidence
  • Heating and temperature control
  • Permanent alignment for calibration stability
  • Easy-to-assemble cell body components (with easy routine maintenance)
  • Chemical resistance to a wide variety of gases
  • Leak-tight, industry-standard sample line connectors
Specifications
DescriptionThermo Scientific™ Antaris™ IGS Gas Cells are optimized for integration with the Antaris IGS gas analyzer.
OptionsOptional heater, Selection of window materials
Product TypeAntaris™ IGS Gas Cells
Sample TypeGases at temperatures up to 185°C and pressures of 1.5 atmospheres
Unit SizeEach