Aluora™ Spatial Amplification Reagent Components
Invitrogen™
Aluora™ Spatial Amplification Reagent Components
Aluora™ Spatial Amplification reagent components enable critical ready-to-use GAM/ GAR and Streptavidin HRP reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityProduct TypeTarget Specificity
AS203HRP200 ReactionsSA HRP Secondary ReagentBiotin
AS100HRP200 ReactionsSpatial Amplification Core Reagent KitNone
AS201HRP200 ReactionsGAM PolyHRP Secondary ReagentMouse
AS202HRP200 ReactionsGAR PolyHRP Secondary ReagentRabbit
A400024508 x 100 ReactionsSpatial Rainbow Kit with Prolong GlassMouse, Rabbit
AS201HRP5PACK1000 ReactionsSecondary ReagentMouse
AS202HRP5PACK1000 ReactionsSecondary ReagentRabbit
AS203HRP5PACK1000 ReactionsSecondary ReagentBiotin
Catalog number AS203HRP
Price (INR)
34,004.00
Each
Quantity:
200 Reactions
Product Type:
SA HRP Secondary Reagent
Target Specificity:
Biotin
Price (INR)
34,004.00
Each
Aluora™ Spatial Amplification reagent components from Invitrogen enable the critical ready-to-use GAM/ GAR and Streptavidin HRP secondary reagent and buffer components for use with the Aluora Spatial Amplification dyes and kits. The secondary antibodies are highly cross absorbed specifically for IgG heavy and light chain for the target species and have a density of HRP that facilitates high fluorescent signal.
Revolutionary signal amplification technology
The Aluora spatial amplification kits represent an advance in signal amplification for spatial proteomics and transcriptomics. They employ proprietary Aluora reagents that react with horseradish peroxidase (HRP) to deposit photostable Aluora dyes onto target proteins, significantly improving the signal-to-background and helping reduce the impact of tissue autofluorescence in commonly imaged wavelengths. This chemistry enables robust multiplexed marker detection even in challenging tissue samples. Aluora kits requires 10–5000X less primary antibody than standard and multiplex IHC with antibodies alone and achieve greater or equal signal intensity.
Aluora spatial amplification reagents are optimized for use with Prolong Glass Antifade mountants and 1 mg/mL Thermo Scientific DAPI nucleic acid stains for multiplex analysis.
Aluora reagents and dyes can be purchased individually for use in IHC and ISH workflows or in pre-configured kits to detect mouse, rabbit, or biotin-labelled antibodies. The Aluora multiplex kits come as dye-only kits configured with 3 to 8 dyes or as the Aluora rainbow kit is configured with 8 Aluora dyes and 4X GAM and 4X GAR secondary detection core reagents for 100 slides in 8-plex.
Features of Aluora spatial amplification reagents include:
  • Signal amplification for enhanced detection sensitivity
  • Comprehensive spatial analysis and imaging capabilities for research applications
  • Fluorescent detection system for multiplexing and quantitative analysis
  • Flexible workflow options that accommodate diverse experimental needs
  • Polymerase HRP (PolyHRP) conjugated secondary antibodies that boost signal for low abundance targets
  • Highly cross-adsorbed secondary antibodies that help ensure target-specific detection and background reduction
  • Enhanced background reduction through proprietary blocking reagents for crisp, clear imaging
Optimized for automated slide staining
A major benefit of Aluora spatial amplification kits is their compatibility with automated slide stainers right out of the box. This enables complete labeling of a tissue slide with an 8-plex Aluora panel and DAPI with walk-away automation sample processing.
Reducing background for cleaner images
Aluora spatial amplification kits include blockers to help reduce endogenous peroxidase activity and autofluorescence. By addressing autofluorescence and background noise, less primary antibody is needed to generate a bright specific signal.
Iterative labeling enables unlimited multiplexing
The covalently bound Aluora dyes are resistant to antibody stripping, remaining brightly fluorescent through subsequent labeling cycles. This enables iterative staining of numerous targets using primary antibodies from the same host species—a major constraint in traditional immunohistochemistry. Any number of directly conjugated primary antibodies can also be introduced after Aluora labeling is complete.
Custom optimization for diverse applications
Although general protocols are provided, each sample requires optimization for optimal results. Tissue preparation and processing methods vary widely and can profoundly impact staining. Careful optimization of the complete workflow is critical to achieve the highest quality multiplexed data from precious samples. Aluora spatial amplification provides a versatile tool to address diverse spatial proteomics applications.
For Research Use Only.
Specifications
ConjugateHRP
Product TypeSA HRP Secondary Reagent
Sample TypeFFPE Tissue
Sufficient ForSpatial Proteomics
Target SpecificityBiotin
For Use With (Application)Spatial Proteomics, Immunohistochemistry (IHC), IHC Staining and Detection, Fluorescent Staining, Immunofluorescence (IF)
Product LineAluora
Quantity200 Reactions
Target SpeciesNone
Unit SizeEach
Contents & Storage
2°C to 8°C and Protect from light