AquaSensors™ AnalogPlus™ Differential pH Sensors
Actual product may vary
Thermo Scientific™
AquaSensors™ AnalogPlus™ Differential pH Sensors
Conduct a wide array of demanding industrial applications with Thermo Scientific™ AquaSensors™ AnalogPlus™ Differential pH Sensors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeDescription
ASAPOR10Differential ORP Sensor1.0 in.
ASAPPH10Differential pH Sensor1.0 in.
Catalog number ASAPOR10
Price (INR)
-
Type:
Differential ORP Sensor
Description:
1.0 in.
Conduct a wide array of demanding industrial applications with Thermo Scientific™ AquaSensors™ AnalogPlus™ Differential pH Sensors. Sensors are available in a choice of materials for a wide range of applications.
  • Replaceable salt bridge to extend sensor life
  • Offered with NTC300 or PT1000 temperature elements
  • Electrode protection options
Specifications
Accuracy0.1% of reading
Description1.0 in.
For Use With (Equipment)AV88 Universal Analyzer, P99 pH/ORP Analyzer
Max. Flow Rate10 ft./sec.
Max. Pressure100psig (at 95°C); 85psig (at 75°C)
Response Time90% in 30 seconds
Temperature (English) Operating23° to 203°F PEEK Sensor Head; 23° to 167°F CPVC Sensor Head
Temperature (Metric) Operating-5° to +95°C (PEEK Sensor Head)/-5° to +75°C (CPVC Sensor Head)
Certifications/ComplianceCE
TypeDifferential ORP Sensor
Unit SizeEach