Oxoid Turbidometer
Product Image
Thermo Scientific™
Oxoid Turbidometer
Standardize inoculum levels and ensure accurate and reliable Antimicrobial Susceptibility Testing (AST) results.
Have Questions?
Catalog NumberQuantity
AU0108A1/Pk.
Catalog number AU0108A
Price (INR)
-
Request A Quote
Quantity:
1/Pk.
Improve reproducibility of inoculum suspension and acquire accurate and reliable Antimicrobial Susceptibility Testing (AST) results using the Thermo Scientific™ Oxoid™ Turbidometer.
Use of a turbidometer allows a simple and quick method to enable complete control of your inoculum to the 0.5 McFarland standard. 
Simply calibrate your Turbidometer with a 0.5 McFarland standard, prepare your inoculum and insert into the test well. A green light will indicate the inoculum level is correct while an orange light indicates the inoculum level is incorrect.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
For Research Use Only. Not for use in diagnostic procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceFor use in USA and Australia
DescriptionOxoid Turbidometer
Quantity1/Pk.
Product TypeMicrobiology Media
Test TypeAntimicrobial Susceptibility
Unit SizeEach