Automated Transmission Sampling Module for Antaris™ II
Automated Transmission Sampling Module for Antaris™ II
Thermo Scientific™
Automated Transmission Sampling Module for Antaris™ II
Thermo Scientific™ Automated Transmission Sampling Module provides quick analysis of liquids, films and paper samples.
Have Questions?
Catalog NumberFor Use With (Equipment)Type
AUTOMATEDTRNSMThermo Scientific Antaris II FT-NIR spectrometerAutomated Transmission Sampling Module
Catalog number AUTOMATEDTRNSM
Price (INR)
-
Request A Quote
For Use With (Equipment):
Thermo Scientific Antaris II FT-NIR spectrometer
Type:
Automated Transmission Sampling Module
Thermo Scientific™ Automated Transmission Sampling Module provides quick analysis of liquids, films and paper samples. The Automated Transmission Sampling Module is a three-position holder that accepts standard-sized cuvettes, culture tubes, vials and self-adhesive cards for heated or unheated transmission analysis.
Features
  • Three-position automated holder
  • Internal dedicated reference position for error-free automated background collections and automated validation
  • Compatible with 4-8mm OD culture tubes
  • Standard-sized cuvettes—0.5 to 10.0mm pathlength
  • Self adhesive transmission cards
  • Optional three-position heated holder
  • Optional external heater
Specifications
DescriptionThe Automated Transmission Sampling Module provides quick analysis of liquids, films and paper samples, with a three-position holder that accepts standard-sized cuvettes, culture tubes, vials and cards for heated or unheated transmission analysis.
Sample TypeLiquids, films and paper samples
For Use With (Equipment)Thermo Scientific Antaris II FT-NIR spectrometer
TypeAutomated Transmission Sampling Module
Unit SizeEach