AutoScript 4
Thermo Scientific™
AutoScript 4
AutoScript is the customization toolkit for electron and Ion microscopy. Built around Python, Autoscript provides researchers the power to automate imaging and associated processing pipes purpose built to solve specific research questions.
Have Questions?
Catalog number AUTOSCRIPT4
Price (INR)
-
Materials discovery often requires advanced techniques for imaging and analysis that cannot be covered in the scope of a general purpose software. AutoScript is the customization toolkit for electron and Ion microscopy. Built around Python, Autoscript provides researchers the power to automate imaging and associated processing pipes purpose built to solve specific research questions.
Key Benefits​
  • Provides a direct link between research needs and microscope automation​
  • Enables improved reproducibility and accuracy
  • Focuses time on the microscope for higher throughput​
Feature Highlights​
  • Python: Autoscript is built upon Python 3.5 and includes a number of pre-installed libraries for scientific computing, data analysis, data visualization, image processing and machine learning.​
  • ​Getting Started: An integrated development environment (IDE) makes it easy to get up and running with autoscript.
  • Scripting Toolbox​: AutoScript 4 comes bundled with a set of commonly needed, prebuilt routines. Not starting from scratch means more time to focus on developing the scripts that define the unique workflow for your research.​