Water Purification Printer
Actual product may vary
Thermo Scientific™
Water Purification Printer
Safely document all measure values and faults with the Thermo Scientific™ Water Purification Printer, compliant with GLP guidelines.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVoltage
AY1137X1110 V
09.2207230 V
Catalog number AY1137X1
Price (INR)
-
Voltage:
110 V
Safely document all measure values and faults with the Thermo Scientific™ Water Purification Printer, compliant with GLP guidelines. This water purification system printer utilizes RS-232 interface and includes date and time in documentation. For use with Thermo Scientific™ Barnstead™ water purification systems.
Utilizes RS-232 interface for safe documentation of all measured values and faults with date and time in compliance with GLP guidelines. Optional accessory for the following Barnstead water purification systems:
  • GenPure™
  • GenPure™ Pro
  • GenPure™ xCAD
  • LabTower™ EDI
  • LabTower™ RO
  • LabTower™ TII
  • Pacific TII™
Specifications
Voltage110 V
For Use With (Equipment)Barnstead, Nanopure, Barnstead RO, Barnstead TII
TypeBarnstead Water Purification Printer
Unit SizeEach