See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
Home
▸
Shop All Products
▸
Cold Storage Products
▸
Refrigerator and Freezer Accessories
▸
CryoClaw
Thermo Scientific™
CryoClaw
Retrieve vials, ampoules and cryoboxes from cryogenic vessels using the Thermo Scientific™ CryoClaw.
Have Questions?
Catalog number
AY509X18
Price (INR)
-
Request A Quote
Retrieve vials, ampoules and cryoboxes from cryogenic vessels using the Thermo Scientific™ CryoClaw.
Rubber suction cups retrieve ampules or boxes that fall from racks or cans into cryogenic vessels
Can also be used to reach overhead items or to safely move radioisotopes without touching containers
Length 41in. (104cm)
Specifications
Description
CryoClaw Ampule Remover
For Use With (Equipment)
All CryoPlus Models
Holds
Vials
Type
CryoClaw