PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
Thermo Scientific™

PTFE-Lined Closed Caps

Use these caps as replacements for our glass environmental sample bottles, jars, jugs and vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck FinishColorProduct Type
B167-3300BL33-400WhiteClosure with Bonded Liner
B267-2414UBB24-414WhiteClosure with Bonded Liner
B167-1000BL110-400WhiteSolid Top
B167-8900BL89-400WhiteClosure with Bonded Liner
B167-3800BL38-400WhiteClosure with Bonded Liner
Catalog number B167-3300BL
Price (INR)
-
Request A Quote
Neck Finish:
33-400
Color:
White
Product Type:
Closure with Bonded Liner

Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.

  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials

Includes:

Cap with bonded liner

Compatible with:

For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.

Specifications
Certification ParametersNone
DescriptionClosed top cap
For Use With (Equipment)Thermo Scientific Glass Environmental Sample Bottles, Jars, Jugs and Vials
LevelUnprocessed
Liner MaterialPTFE
Neck Finish33-400
No. per Case24
Certifications/ComplianceManufactured under an ISO certified quality management system.
ColorWhite
MaterialPolypropylene
Product TypeClosure with Bonded Liner
ShapeRound
Unit SizeCase of 24