PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
PTFE-Lined Closed Caps
Thermo Scientific™

PTFE-Lined Closed Caps

Use these caps as replacements for our glass environmental sample bottles, jars, jugs and vials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeck FinishColorProduct Type
B167-8900BL89-400WhiteClosure with Bonded Liner
B267-2414UBB24-414WhiteClosure with Bonded Liner
B167-1000BL110-400WhiteSolid Top
B167-3300BL33-400WhiteClosure with Bonded Liner
B167-3800BL38-400WhiteClosure with Bonded Liner
Catalog number B167-8900BL
Price (INR)
-
Neck Finish:
89-400
Color:
White
Product Type:
Closure with Bonded Liner

Thermo Scientific™ PTFE-Lined Closed Caps are replacement caps for Thermo Scientific glass environmental sample bottles, jars, jugs and vials. The same high quality PTFE-lined caps used on our Thermo Scientific glass environmental sample containers. The PTFE liner is bonded to the cap so it won't fall out or become separated from the cap.

  • White polypropylene closed top cap with bonded PTFE liner
  • 24-414 cap has a PTFE-faced silicone liner for added cushion and an extra secure seal when used with bottles and VOA vials

Includes:

Cap with bonded liner

Compatible with:

For use with Thermo Scientific glass environmental sample bottles, jars, jugs and vials.

Specifications
Certification ParametersNone
DescriptionClosed top cap
LevelUnprocessed
Liner MaterialPTFE
Neck Finish89-400
No. per Case24
Certifications/ComplianceManufactured under an ISO certified quality management system.
ColorWhite
MaterialPolypropylene
Product TypeClosure with Bonded Liner
ShapeRound
Unit SizeCase of 24