KingFisher™ Apex Dx Magnetic Heads
Thermo Scientific™
KingFisher™ Apex Dx Magnetic Heads
The Thermo Scientific KingFisher Apex Dx Purification Instrument allows for extraction of nucleic acids from human specimens with a small, compact footprint making it ideal for use in diagnostic and development workflows.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
B24079930Apex Dx 96 Deep Well Head
B24079940Apex Dx 24 Combi Head
B24079920Apex Dx 96 Combi Head
B24079910Apex Dx 96 PCR Head
Catalog number B24079930
Price (INR)
-
Format:
Apex Dx 96 Deep Well Head
KingFisher Apex Dx Magnetic Heads are for use with the KingFisher Apex Dx Purification Instrument. The instrument accommodates two interchangeable magnetic heads (it comes with one included). The interchangeable heads and heating blocks provide flexibility in the extraction of nucleic acids, proteins, and cells with a small, compact footprint, making the instrument ideal for use in diagnostic and development workflows.
With its intuitive software, automation capabilities, and revolutionary magnetic particle separation technology, the KingFisher Apex Dx Purification Instrument provides high quality, exceptional reliability, and outstanding run-to-run reproducibility for clinicians and assay developers seeking an IVD sample preparation solution for their diagnostic workflow.
For in vitro diagnostic use.
Specifications
DisposableReusable
Quantity1 head
FormatApex Dx 96 Deep Well Head
Product TypeNucleic Acid Extraction and Cell Separation Instrument
Sample TypeViral Samples
Unit SizeEach