DRI Tramadol Calibrator Set
Thermo Scientific™

DRI Tramadol Calibrator Set

Ensure accurate calibration with the Thermo Scientific DRI Tramadol High and Low Control Set, designed for urine screening with the Thermo Scientfic DRI Tramadol Drugs of Abuse Assay. The control set includes a high and low control targeted at 75 ng/mL and 125 ng/mL respectively, to validate the calibration curve by providing positive and negative results.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityConcentrationType
B400003522 x 10 mL100 ng/mLCutoff Calibrator
B400003532 x 10 mL0 ng/mLNegative Calibrator
B400003665 x 10 mL0, 100, 200, 500 and 1000 ng/mLCalibrator Set
Catalog number B40000352
Price (INR)
-
Quantity:
2 x 10 mL
Concentration:
100 ng/mL
Type:
Cutoff Calibrator

Ensure accurate calibration with the Thermo Scientific DRI Tramadol High and Low Control Set, designed for urine screening with the Thermo Scientfic DRI Tramadol Drugs of Abuse Assay. The control set includes a high and low control targeted at 75 ng/mL and 125 ng/mL respectively, to validate the calibration curve by providing positive and negative results.

  • Specifically detect tramadol and its primary metabolites O-Desmethyltramadol and N-Desmethyltramadol at physiologically relevant concentrations in human urine
  • Low cross-reactivity to non-tramadol compounds
  • High sensitivity with a cutoff concentration of 100 ng/mL
  • Superior assay precision
  • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to Use
For In Vitro Diagnostic Use.  
Specifications
Certifications/ComplianceFDA Cleared
Concentration100 ng/mL
For Use With (Equipment)Automated Chemistry Analyzers
Quantity2 x 10 mL
TypeCutoff Calibrator
Detectable AnalytesTramadol
Sample TypeUrine
Unit SizeEach