Replacement Viper™ Capillaries for Dionex™ Inuvion™ IC System, PEEK
Replacement Viper™ Capillaries for Dionex™ Inuvion™ IC System, PEEK
Thermo Scientific™

Replacement Viper™ Capillaries for Dionex™ Inuvion™ IC System, PEEK

Ensure optimal separations and maximize ion chromatography (IC) performance with virtually-zero-dead-volume connections.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberOverall Length (Metric)
B51000237203 mm
B5100023376 mm
B51000230140 mm
Catalog number B51000237
Price (INR)
-
Request A Quote
Overall Length (Metric):
203 mm

Ensure optimal separations and maximize ion chromatography (IC) performance with virtually-zero-dead-volume connections. Thermo Scientific™ Viper™ Fingertight Fittings with biocompatible PEEK capillaries are easy to use and enable fast, tool-free fluidic setup of IC systems. These fittings are compatible with virtually any column and valve type and suitable for fluidic connections with pressures up to 345 bar (5,000 psi).

The ferrule-free Dionex IC PEEK Viper design reduces dead volume of fluidic connections to virtually zero by sealing at the tip of the tubing. Dionex IC PEEK Viper fittings require much less torque to tighten than traditional PEEK fittings. The effort required to create a seal for 5000 psi (34 MPa) of pressure is similar to the effort needed to turn a dial on a combination lock.

Specifications
DescriptionPEEK Viper Fitting, 0.065 x 203 mm
For Use WithDionex Inuvion IC System
Diameter (Metric) Inner65 μm
MaterialPEEK
Overall Length (Metric)203 mm
Unit SizeEach