5 in. (127 mm) PharMed™ BPT Replacement Tubing for Dionex™ Inuvion™ Integrated Regenerant Pump
Thermo Scientific™
5 in. (127 mm) PharMed™ BPT Replacement Tubing for Dionex™ Inuvion™ Integrated Regenerant Pump
Ensure the optimum flow of your Thermo Scientific™ Dionex™ Inuvion™ suppressor regenerant solution with replacement single-channel peristaltic pump tubing.
Have Questions?
Catalog NumberFor Use With
B51001284Dionex Inuvion IC System
Catalog number B51001284
Price (INR)
-
For Use With:
Dionex Inuvion IC System
Ensure the optimum flow of your Thermo Scientific™ Dionex™ Inuvion™ suppressor regenerant solution with replacement single-channel peristaltic pump tubing.
Optional accessory for all Dionex Inuvion IC system configurations.
  • For use with Dionex Inuvion Integrated Regenerant Pump.
Specifications
Description5 in. (127 mm) PharMed™ BPT Replacement Tubing for Dionex Inuvion Integrated Regenerant Pump
For Use WithDionex Inuvion IC System
TypePeristaltic Tubing
Unit SizeEach