Dionex™ Inuvion™ Preventive Maintenance Kit
Dionex™ Inuvion™ Preventive Maintenance Kit
Dionex™ Inuvion™ Preventive Maintenance Kit
Dionex™ Inuvion™ Preventive Maintenance Kit
Thermo Scientific™
Dionex™ Inuvion™ Preventive Maintenance Kit
The Dionex™ Inuvion™ Preventive Maintenance Kit is designed to meet the preventative maintenance needs of your Dionex hardware.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51001547Dionex Inuvion IC System
Catalog number B51001547
Price (INR)
-
For Use With (Equipment):
Dionex Inuvion IC System
Safeguard your lab productivity and protect your investment by performing routine maintenance on your Thermo Scientific™ Dionex™ Inuvion™ IC system with the help of this preventive maintenance kit. Preventive care is critical to the life and availability of your IC system and is recommended on an annual basis. Without regular maintenance, unattended, lingering problems can decrease instrument performance and lead to unnecessary and unplanned service costs.
The Dionex Inuvion preventive maintenance kit includes the following items:
  • Large seal wash O-ring (1)
  • Small seal wash O-ring (1)
  • Main pump piston seal (2)
  • Rear seal wash seal (2)
  • Check valve cartridge (2)
  • 6-port injection valve rebuild kit (1)
  • Eluent end-line filter (1)
  • Reminder label for next preventative maintenance
Specifications
DescriptionPreventive Maintenance kit for Dionex Inuvion IC systems
For Use With (Equipment)Dionex Inuvion IC System
Unit SizeEach