OptiSpray Ion Source
OptiSpray Ion Source
OptiSpray Ion Source
OptiSpray Ion Source
OptiSpray Ion Source
Thermo Scientific™
OptiSpray Ion Source
Reliably achieve maximum ESI performance for proteomics, metabolomics, and lipidomics experiments across LC-MS systems, columns, and users of all experience levels with the Thermo Scientific™ OptiSpray™ Ion source.
NOT ORDERABLE
Have Questions?
Catalog number B51004132
Price (INR)
-
Take your research to the next level with the Thermo Scientific™ OptiSpray™ Ion Source, an intelligent electrospray ionization interface that dramatically simplifies the acquisition of high-quality, reproducible nano and capillary LC-MS data. Compatible with our leading mass spectrometer portfolio and the Thermo Scientific™ Vanquish™ Neo UHPLC system, the OptiSpray ion source combines automated spray optimization with cartridge-based nano and capillary columns to deliver high-sensitivity, quantitative accuracy and precision, and maximum robustness for discovery and targeted assays.
Along with Thermo Scientific™ OptiSpray™ Cartridge columns, the OptiSpray ion source provides:
Performance for deep coverage with high precision and accuracy
Optimized fluidics, integrated temperature and sheath gas control, and automated spray positioning yield reproducible, high-quality results when used with or without the FAIMS interface.
Simplicity 
for users of all experience levels 
Plug-and-play cartridge installation and automated spray optimization virtually eliminate manual setup of nano and capillary columns. On-board memory stores cartridge properties, usage data, and previous source settings to quickly achieve optimal ESI performance, every time.
Reliability across columns, users, and LC-MS systems
Enhanced robustness with an inactive spray position between injections for a cleaner MS system while exchangeable emitters extend column lifetime. Automated spray positioning, cartridge memory, and integrated nebulization gas boost run-to-run and intercolumn result reproducibility.
Specifications
For Use With (Application)LC-MS
For Use With (Equipment)OptiSpray Cartridge Columns
TypeOptiSpray Ion Source
Unit SizeEach