EXTREVA™ ASE™ Solvent Bottle Tubing and Caps for PFAS Analysis
This solvent bottle tubing and these caps should be used with the Thermo Scientific™ EXTREVA™ ASE™ Accelerated Solvent Extractor to provide PFAS solvents to the instrument.
Have Questions?
Catalog Number
For Use With (Application)
B51004605
Sample Extraction, PFAS Analysis
Catalog number B51004605
Price (INR)
-
For Use With (Application):
Sample Extraction, PFAS Analysis
Use this PEEK tubing and these solvent bottle caps with the extraction solvent bottles of the EXTREVA ASE system for PFAS analysis.
For use with the EXTREVA ASE system
Specifications
For Use With (Application)Sample Extraction, PFAS Analysis