Cindion™ Solids and Liquids Kits
Cindion™ Solids and Liquids Kits
Cindion™ Solids and Liquids Kits
Thermo Scientific™
Cindion™ Solids and Liquids Kits
Use these kits to set up your Thermo Scientific™ Cindion™ Combustion Ion Chromatography System for solid- or liquid-sample analysis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
B51006426Liquids Kit
B51006427Solids Kit
Catalog number B51006426
Price (INR)
-
Type:
Liquids Kit
Use the Thermo Scientific™ Cindion™ Solids Kit or the Thermo Scientific™ Cindion™ Liquids Kit to set up your Cindion Combustion Ion Chromatography (C-IC) system for solid- or liquid-sample analysis, respectively.
Cindion Solids Kit
This kit includes a gripper, tray, boat, and cups for solid samples. Pair this kit with other key components of the Cindion C-IC system to perform combustion IC analysis for solid samples. Configurations for solid samples include:
  • Solid-sample analysis
  • Solid-sample analysis with US EPA Method 1621 compatibility
Cindion Liquids Kit
This kit includes a syringe holder, tray, boat, vials, syringe, and needles for liquid samples. Pair this kit with other key components of the Cindion C-IC system to perform combustion IC analysis for liquid samples.
Specifications
For Use With (Application)Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System
IncludesSyringe Holder, Tray, Boat, Vials, Syringe, Needles for Liquid Samples
TypeLiquids Kit
Unit SizeEach