Cindion™ Autosampler
Thermo Scientific™
Cindion™ Autosampler
Automate sample injection of solid or liquid samples for combustion ion chromatography analysis with the Thermo Scientific™ Cindion™ Autosampler.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51006429Cindion Combustion IC System
Catalog number B51006429
Price (INR)
-
For Use With (Equipment):
Cindion Combustion IC System
Automate sample delivery of solid or liquid samples for combustion IC (C-IC) analysis with the Cindion Autosampler. This versatile autosampler allows you to easily switch between solid and liquid samples for C-IC analysis with the Thermo Scientific™ Cindion™ Combustion IC System without the need for tools.
Cindion Autosampler features
  • Load up to 45 solid or 120 liquid samples
  • Small footprint - integrates with the Thermo Scientific™ Cindion™ Combustion/Absorption Module
Cindion Combustion IC System configurations
Pair the Cindion Autosampler with other key components of the Cindion C-IC System to perform combustion IC analysis for solid or liquid samples. Configurations include:
  • Solid-sample analysis
  • Solid-sample analysis with US EPA Method 1621 compatibility
  • Liquid-sample analysis
Specifications
Capacity120 Liquid Samples or 45 Solid Samples
Dimensions (D x W x H)31 cm x 63 cm x 43 cm
For Use With (Application)Combustion IC for Solid Samples, Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System
Weight (English)17.3 lb
Weight (Metric)7.8 kg
TypeAutosampler
Unit SizeEach
Frequently asked questions (FAQs)
How has the Cindion C-IC system achieved a relatively small footprint?
By combining the combustion oven, absorption module, and autosampler into one integrated instrument, the Cindion C-IC system minimizes bench space requirements. The compact Z-fold combustion tube also helps by offering efficient combustion within a smaller oven.