Cindion™ Glass Connector Outlet
Thermo Scientific™
Cindion™ Glass Connector Outlet
Use the Thermo Scientific™ Cindion™ Glass Connector Outlet to facilitate sample combustion and absorption for combustion IC analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Catalog NumberFor Use With (Equipment)
B51006434Cindion Combustion IC System
Catalog number B51006434
Price (INR)
-
For Use With (Equipment):
Cindion Combustion IC System
This glass connector outlet connects the combustion tube to the tubing that feeds into the absorber tube.
  • This connector outlet is an accessory for all Cindion Combustion IC System configurations.
Specifications
For Use With (Application)Combustion Ion Chromatography
For Use With (Equipment)Cindion Combustion IC System
MaterialGlass
TypeCombustion/Absorption Module Accessory
Unit SizeEach