Cindion™ Solid and Liquid Boats
Thermo Scientific™
Cindion™ Solid and Liquid Boats
Thermo Scientific™ Cindion™ Solid and Liquid Boats boats hold samples for use in combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
B51006443Solids Boat
B51006445Liquids Boat
Catalog number B51006443
Price (INR)
-
Type:
Solids Boat
Use these quartz sample boats to hold samples for insertion into the combustion tube for combustion-ion chromatography (C-IC) analysis. These boats are available for solid or liquid samples.
These boats are compatible with solid and liquid sample configurations of the Cindion Combustion IC System.
  • Boat is introduced into the combustion oven at a programmed rate
Specifications
Dimensions (D x W x H)2 cm x 16 cm x 2 cm
For Use With (Application)Combustion IC for Solid Samples
For Use With (Equipment)Cindion Combustion IC System
MaterialQuartz
TypeSolids Boat
Unit SizeEach