Cindion™ Sample Cups
Thermo Scientific™
Cindion™ Sample Cups
Thermo Scientific™ Cindion™ Sample Cups hold solid or liquid samples for combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialQuantity
B51006446Quartz1
B51006447Quartz45
B51006448Ceramic3
Catalog number B51006446
Price (INR)
-
Material:
Quartz
Quantity:
1
Use Cindion Sample Cups to hold solid or liquid samples for combustion-ion chromatography (C-IC) analysis. These sample cups are available in quartz or ceramic.
These cups are compatible with the solid sample configuration of the Cindion Combustion IC System.
  • Available as quartz cups or ceramic cups
Specifications
For Use With (Application)Combustion IC
For Use With (Equipment)Cindion Combustion IC System
MaterialQuartz
Quantity1
TypeSample Cups
Unit SizeEach