Use Thermo Scientific™ Cindion™ Septa to seal the sample port for liquid-sample injections during combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Catalog Number
For Use With (Equipment)
B51006449
Cindion Combustion IC System
Catalog number B51006449
Price (INR)
-
For Use With (Equipment):
Cindion Combustion IC System
Seal the sample port for liquid-sample injections for combustion ion chromatography (C-IC) analysis using Cindion Septa.
These septa are used in the liquid-sample configuration of the Cindion Combustion IC System.
Available in pack of 10
Specifications
Diameter12 mm
For Use With (Application)Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System