Pair Thermo Scientific™ Cindion™ Syringes with Thermo Scientific™ Cindion™ Blunt Point Needles to withdraw and dispense liquid samples for combustion ion chromatography analysis using the Thermo Scientific™ Cindion™ Combustion IC System.
Have Questions?
Change view
Catalog Number
Volume (Metric)
B51008999
500 μL
B51008996
10 μL
B51008997
25 μL
B51008998
100 μL
B51008977
250 μL
5 Options
Catalog number B51008999
Price (INR)
-
Volume (Metric):
500 μL
Use these syringes to withdraw and dispense liquid samples for combustion-ion chromatography (C-IC) analysis using the Cindion C-IC System. They come in a variety of volumes and attach to the autosampler syringe holder plate to dispense samples into the liquids boat prior to C-IC analysis.
These syringes are for use with the liquid-sample configuration of the Cindion Combustion IC System
Available in 5 volumes: 10, 25, 100, 250, and 500 μL
Removable needle; needles not included with syringe
Use with Cindion Blunt Point Needles
Specifications
Dimensions (D x W x H)10 x 1 x 1 cm
For Use With (Application)Combustion IC for Liquid Samples
For Use With (Equipment)Cindion Combustion IC System