Alpha-Bungarotoxin Conjugates
Invitrogen™

Alpha-Bungarotoxin Conjugates

Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLabel or Dye
B56130Alexa Fluor™ Plus 405
B13422Alexa Fluor™ 488
B35451Alexa Fluor™ 555
B13423Alexa Fluor™ 594
B35450Alexa Fluor™ 647
B1196Biotin-XX
T1175Tetramethylrhodamine
B1601Unlabeled
Catalog number B56130
Price (INR)
38,172.00
キャンペーン価格
Ends: 31-Mar-2026
63,620.00
Save 25,448.00 (40%)
Each
Label or Dye:
Alexa Fluor™ Plus 405
Price (INR)
38,172.00
キャンペーン価格
Ends: 31-Mar-2026
63,620.00
Save 25,448.00 (40%)
Each
Thermo Fisher Scientific offers a broad selection of Invitrogen alpha-bungarotoxin conjugates, including Alexa Fluor and Alexa Fluor Plus conjugates. Alpha-bungarotoxin, a 74-amino acid peptide extracted from Bungarus multicinctus venom, binds with high affinity to the alpha subunit of the nicotinic acetylcholine receptor (AChR) of neuromuscular junctions.

Our bright and photostable Alexa Fluor 555 alpha-bungarotoxin is a superior choice of probe for visualizing this receptor. Fluorescent alpha-bungarotoxin conjugates can be used to facilitate identification of nicotinic AChRs and localization of neuromuscular junctions. Conjugated alpha-bungarotoxin has been used in variety of applications, including immunofluorescence (IF), immunohistochemistry (IHC), and flow cytometry (FC).

Specifications
ColorViolet
Label or DyeAlexa Fluor™ Plus 405
Quantity100 μg
Excitation/Emission411/431 nm
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store at ≤-20°C, protect from light. Avoid freeze-thaw cycles.