The 10X FastDigest Green Buffer is a proprietary digestion buffer which supports 100% activity of all FastDigest restriction enzymes. DNA/RNARead more
Have Questions?
Catalog Number
Quantity
B72
5 x 1 mL
Catalog number B72
Price (INR)
2,760.00
Each
Quantity:
5 x 1 mL
Request bulk or custom format
Price (INR)
2,760.00
Each
The 10X FastDigest Green Buffer is a proprietary digestion buffer which supports 100% activity of all FastDigest restriction enzymes. DNA/RNA modifying enzymes, such as ligases, phosphatases, kinases and mesophilic DNA polymerases also have 100% activity in FastDigest Green Buffer. Therefore, enzymes used in downstream applications can be directly added to the FastDigest reaction mix
Benefits: • Double and multiple digestion in a single buffer with any combination of enzymes. • No sequential digestions or buffer changes needed. • Allows direct loading of reaction mixture on gels. • Saves pipetting time and eliminates the need for DNA loading dyes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialDigestion Buffer
Concentration10X
For Use With (Application)Traditional Cloning
Product LineFastDigest
Quantity5 x 1 mL
Unit SizeEach
Frequently asked questions (FAQs)
Can I store the FastDigest Buffers at 4 degrees C?
The FastDigest Buffers can be stored at 4 degrees C for at least 6 months. For long-term storage until their expiration, we recommend storing the FastDigest and FastDigest Green Buffers at -20 degrees C.
How I can transform or calculate reaction numbers in the unit of FastDigest enzymes?
The concentration of FastDigest enzymes is proprietary information. The FastDigest enzyme activity is measured by FastDigest unit (FDU): 1 µL of enzyme (1 FDU) cleaves 1 µg of DNA substrate in 5-15 min at 37 degrees C in 20 µL of 1X FastDigest buffer.
Are the FastDigest and FastDigest Green Buffer available separately?
Yes, the FastDigest Buffer (Cat. No. B64) and the FastDigest Green Buffer (Cat. No. B71) can be purchased separately.
What is the difference between FastDigest Buffer and FastDigest Green Buffer?
The FastDigest Green Buffer offers the same performance as the colorless FastDigest Buffer but enables direct loading of the reaction mixture on gels. The 10X FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading. The blue dye migrates with 3-5 kb DNA fragments in a 1% agarose gel and has an excitation peak of 424 nm. The yellow dye migrates faster than 10 bp DNA fragments in a 1% agarose gel and has an excitation peak of 615 nm.