Product Image
Gibco™

Sf21 Cells in Grace's

Gibco™ Sf21 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins. The cellsRead more
Have Questions?
Catalog NumberQuantity
B82101
also known as B821-01
1 mL
Catalog number B82101
also known as B821-01
Price (INR)
56,241.00
Online offer
Ends: 31-Dec-2026
83,320.00
Save 27,079.00 (33%)
Each
Quantity:
1 mL
Price (INR)
56,241.00
Online offer
Ends: 31-Dec-2026
83,320.00
Save 27,079.00 (33%)
Each
Gibco™ Sf21 cells are commonly used to isolate and propagate recombinant baculoviral stocks and to produce recombinant proteins. The cells originated at the USDA Insect Pathology Laboratory, where they were derived from the pupal ovarian tissue of the fall army worm, Spodoptera frugiperda. Gibco™ Sf21 cells (frozen in Grace's Insect Medium) feature:
• Recombinant protein expression from a variety of expression systems
• Good growth in adherent or suspension culture
• Quality and performance testing

Recombinant protein expression from a variety of expression systems
High levels of protein expression in Sf21 cells can be obtained using either the BaculoDirect™ Baculovirus Expression System, the Bac-to-Bac™ Baculovirus Expression System, or the InsectDirect™ Expression System.

Good growth in adherent or suspension culture
Protocols for adherent or suspension growth in Gibco™ Supplemented Grace's Insect Medium are available in the product manual. Cultures can easily be transferred between the two conditions for ease of workflow.

Quality and performance testing
Each lot of Gibco™ Sf21 cells is tested for cell growth and viability post-recovery from cryopreservation.

Caution: Handle as potentially biohazardous material under at least Biosafety Level 2 containment. This product contains Dimethyl Sulfoxide (DMSO), a hazardous material. Review the Material Safety Data Sheet before handling.
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
Specifications
Product TypeInsect Cells
Quantity1 mL
Cell LineSf21
Cell TypeInsect Cells
SpeciesS. frugiperda
Unit SizeEach
Contents & Storage
Storage conditions: Liquid nitrogen (vapor phase)
Shipping conditions: Frozen

Frequently asked questions (FAQs)

What is the procedure to thaw frozen insect cells?

The following protocol describes a general procedure for thawing cryopreserved cells. For detailed protocols, always refer to the cell-specific product insert.

1. Remove the cryovial containing the frozen cells from liquid nitrogen storage and immediately place it into a 37°C water bath.
2. Quickly thaw the cells (< 1 minute) by gently swirling the vial in the 37°C water bath until there is just a small bit of ice left in the vial.
3. Transfer the vial into a laminar flow hood. Before opening, wipe the outside of the vial with 70% ethanol.
4. Transfer the desired amount of pre-warmed complete growth medium appropriate for your cell line dropwise into the centrifuge tube containing the thawed cells.
5. Centrifuge the cell suspension at approximately 200 x g for 5-10 minutes. The actual centrifugation speed and duration varies depending on the cell type.
6. After the centrifugation, check the clarity of supernatant and visibility of a complete pellet. Aseptically decant the supernatant without disturbing the cell pellet.
7. Gently resuspend the cells in complete growth medium, and transfer them into the appropriate culture vessel and into the recommended culture environment.

Note: The appropriate flask size depends on the number of cells frozen in the cryovial, and the culture environment varies based on the cell and media type.

Why does the Insect cell line manual state: "Cells should be maintained at 27 degrees C in a non-humidified environment."

Insect cells do not require CO2 or high humidity to grow, they can grow in a lab drawer at room temperature. We recommend this so people don't waste CO2 and other resources necessary for maintaining a tissue culture incubator. It should be noted, however, that the cells will grow in a humidified incubator.

What methods can be used to detach tightly adherent Sf9 and Sf21 cells cultured in Grace's media supplemented with 10% heat-inactivated FBS, Sf-900 II SFM, or Sf-900 III SFM?

Sf9 and Sf21 cells should be lightly adherent cells. However, there are some Sf9 and Sf21 cells that attach to culture vessels very tightly. The use of enzymes such as trypsin, collagenase, hyaluronidase, TrypLE Express, and TrypLE Select have been tried without success for passaging cells. The main problem is that the cells do not attach well after having dissociated with the enzymes.

The best method to use is to culture cells in a T-flask. Close cap tightly and hold flask with cap pointing towards the ceiling. Hit the bottom of the flask over a counter 2-3 times with medium force. Cell detachment may be 60-80% and not 100%. This will allow for detachment of enough cells for passaging. If tapping the flask over the counter is performed with too harsh of a force or too many times, cell viability will be greatly affected.

If possible, we recommend that you culture cells in suspension conditions. Cells in suspension cultures can be passaged directly into adherent conditions when needed. The culture of cells in suspension conditions will allow for higher cell densities as cell growth is not limited to the surface area.

How can I concentrate my insect cells to increase the cell density?

If the cell density is too low and the cells have been in culture for 4-5 days, we recommend concentrating the cells by centrifuging them at 100 X g for 5 minutes and resuspending them in fresh medium. Cells should not be left in the same medium for more than 4-5 days as nutrients in the medium will have been used up by the cells in that period, and the medium itself degraded due to prolonged exposure to warm temperatures. Cells should also be centrifuged and concentrated if a lot of cell debris is observed in culture.

What are the main differences between insect cell culture and mammalian cell culture?

Insect cells are much more fragile than a lot of mammalian cell lines. They suffer much more damage than mammalian cells from overgrowth and over-splitting. Never let cells go above 8 x 10E6 cells/mL or grow at densities less than 0.5 x 10E6 cells/mL in suspension. Insect cells require a little more osmotic pressure than mammalian cells (340 µOsM). Insect cells use a lot of O2, especially during protein expression. Insect cell culture media is more acidic than mammalian media (pH 6.0-6.4). The insect cell culture media is phosphate buffer based. Therefore, no CO2 is needed to maintain the pH.

Citations & References (9)

Citations & References
Abstract
The enzymological basis for resistance of herpesvirus DNA polymerase mutants to acyclovir: relationship to the structure of alpha-like DNA polymerases.
Authors:Huang L, Ishii KK, Zuccola H, Gehring AM, Hwang CB, Hogle J, Coen DM
Journal:Proc Natl Acad Sci U S A
PubMed ID:9892653
'Acyclovir (ACV), like many antiviral drugs, is a nucleoside analog. In vitro, ACV triphosphate inhibits herpesvirus DNA polymerase by means of binding, incorporation into primer/template, and dead-end complex formation in the presence of the next deoxynucleoside triphosphate. However, it is not known whether this mechanism operates in vivo. To address ... More
15-Lipoxygenase metabolism of 2-arachidonylglycerol. Generation of a peroxisome proliferator-activated receptor alpha agonist.
Authors: Kozak Kevin R; Gupta Rajnish A; Moody John S; Ji Chuan; Boeglin William E; DuBois Raymond N; Brash Alan R; Marnett Lawrence J;
Journal:J Biol Chem
PubMed ID:11956198
'The recent demonstrations that cyclooxygenase-2 and leukocyte-type 12-lipoxygenase (LOX) efficiently oxygenate 2-arachidonylglycerol (2-AG) prompted an investigation into related oxygenases capable of metabolizing this endogenous cannabinoid receptor ligand. We evaluated the ability of six LOXs to catalyze the hydroperoxidation of 2-AG. Soybean 15-LOX, rabbit reticulocyte 15-LOX, human 15-LOX-1, and human 15-LOX-2 ... More
Structure of the reovirus membrane-penetration protein, Mu1, in a complex with is protector protein, Sigma3.
Authors: Liemann Susanne; Chandran Kartik; Baker Timothy S; Nibert Max L; Harrison Stephen C;
Journal:Cell
PubMed ID:11832217
Cell entry by nonenveloped animal viruses requires membrane penetration without membrane fusion. The reovirus penetration agent is the outer-capsid protein, Mu1. The structure of Mu1, complexed with its  ... More
Discovery of a small molecule insulin mimetic with antidiabetic activity in mice [see comments]
Authors:Zhang B, Salituro G, Szalkowski D, Li Z, Zhang Y, Royo I, Vilella D, Diez MT, Pelaez F, Ruby C, Kendall RL, Mao X, Griffin P, Calaycay J, Zierath JR, Heck JV, Smith RG, Moller DE
Journal:Science
PubMed ID:10320380
Insulin elicits a spectrum of biological responses by binding to its cell surface receptor. In a screen for small molecules that activate the human insulin receptor tyrosine kinase, a nonpeptidyl fungal metabolite (L-783,281) was identified that acted as an insulin mimetic in several biochemical and cellular assays. The compound was ... More
Proteolytic activity of human osteoclast cathepsin K. Expression, purification, activation, and substrate identification.
Authors:Bossard MJ, Tomaszek TA, Thompson SK, Amegadzie BY, Hanning CR, Jones C, Kurdyla JT, McNulty DE, Drake FH, Gowen M, Levy MA
Journal:J Biol Chem
PubMed ID:8647860
Human cathepsin K is a recently identified protein with high primary sequence homology to members of the papain cysteine protease superfamily including cathepsins S, L, and B and is selectively expressed in osteoclasts (Drake, F.H., Dodds, R., James I., Connor J., Debouck, C., Richardson, S., Lee, E., Rieman, D., Barthlow, ... More